Order Entry
Switzerland
ContactUsLinkComponent
 
 
1,8-Diaminonaphthalene for synthesis, Sigma-Aldrich®
 :  Merck
 
 
1,8-Diaminonaphthalene for synthesis, Sigma-Aldrich®
 :  Merck

 

  • Solubility:
    (20 °C) slightly soluble
  • Melting Point:
    61 - 65 °C
  • Molar Mass:
    158.2 g/mol
  • Boiling Point:
    205 °C (15.6 hPa)
  • Vapor Pressure:
    195 °C

 

Frequently Bought Together