
Ammonium cerium(IV) sulphate dihydrate ≥94% ACS
Supplier: Thermo Fisher Scientific
Certificates
About this item
CAS Number:10378-47-9
:
Formula:(NH₄)₂Ce(SO₄)₄·2H₂O
MDL Number:MFCD00149417
:632.55 g/mol
Melting Pt:130 °C
EINECS:231-567-4
:GHS07
Storage Temperature:Ambient
Related Information
Frequently Bought Together