Order Entry
Switzerland
ContactUsLinkComponent
Varicella zoster virus IgG ELISA kit
Varicella zoster virus IgG ELISA kit
Catalog # ABNOKA1456
Supplier:  Abnova
CAS Number:  
Varicella zoster virus IgG ELISA kit
Catalog # ABNOKA1456
Supplier:  Abnova
CAS Number:  

Specifications

  • Pk:
    1 KIT
  • Target protein:
    Varicella zoster virus IgG
  • Description:
    ELISA kit, 100 µl
  • Sample type:
    Cerebrospinal fluid, serum
  • Storage temperature:
    4 °C
  • Application:
    Quantification

Specifications

About this item

Varicella zoster virus IgG ELISA Kit is a solid phase enzyme immunoassay for the quantitative measurement of IgG class autoantibodies against Varicella zoster virus.

  • HRP-conjugate
  • Colorimetric detection
Caution: For research use only.