Holmium(III) oxide 99.9% (REO, rare earth oxide basis) -325 mesh
Supplier: Thermo Fisher Scientific
Certificates
About this item
Synonyms: Diholmium trioxide, Holmium oxide
CAS Number:12055-62-8
:
Formula:Ho₂O₃
MDL Number:MFCD00011053
Boiling Pt:3900 °C (760 mmHg)
Molecular Weight:377.86 g/mol
Melting Pt:2330 °C
Storage Temperature:Ambient
EINECS:235-015-3
Specifications
- Environmentally Preferable:
Specifications
Specification Test Results
- Appearance:Yellow to beige to light orange Powder
- Assay:≥99.85 % (Ho2O3) TREO base
- Particle size:≤325 mesh
- Loss on ignition:≤1.0 % (1000°C)
Specification Test Results
Frequently Bought Together