Order Entry
Switzerland
Orders LinkContactUsLinkComponent
7-Hydroxyindole

7-Hydroxyindole

Glass bottle for solids, 1 g
Catalog # MOLE18308008-1G
Marketsource
Supplier:   Molekula
7-Hydroxyindole
Catalog # MOLE18308008-1G
Supplier:  Molekula
Marketsource
CAS Number:  2380-84-9
7-Hydroxyindole

Glass bottle for solids, 1 g

CAS Number:  2380-84-9
Price
Inquire For Price
Log in for availability
1 * 1 g
Product Details & Documents
  • CAS Number:2380-84-9
  • Formula:C₈H₇NO
  • MDL Number:MFCD00152102
  • Molecular Weight:133.15 g/mol
Specifications

Specifications

  • Pack type:
    Glass bottle for solids
  • Pk:
    1 g

Specifications

Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.