Order Entry
Switzerland
ContactUsLinkComponent
Human Recombinant GRObeta (from E. coli)
Human Recombinant GRObeta (from E. coli)
Certificates
undefined
Human Recombinant GRObeta (from E. coli)

Specifications

  • Conjugation:
    Unconjugated
  • Protein function:
    Cytokine
  • Protein/peptide type:
    Recombinant
  • Source:
    E. coli
  • Species:
    Human
  • Storage conditions:
    –20 °C
  • Endotoxin content:
    Endotoxin LAL (EU/µg) ≤1
  • Biological activity:
    Bioactivity: No biological activity data is available at this time
  • Gene ID:
    P19875
  • Reconstitution instructions:
    Sterile water at 0.1 mg/ml
  • Endotoxin-free:
    N
  • Carrier-free:
    Y
  • Protease-free:
    N
  • Animal-free:
    N
  • Protein synonyms:
    CXCL2|GRO2|MIP-2α|MGSAβ
  • Protein/peptide name:
    GRO-beta
  • Purity:
    ≥95%
  • Molecular weight:
    7,9 kDa
  • Sequence:
    APLATELRCQ CLQTLQGIHL KNIQSVKVKS PGPHCAQTEV IATLKNGQKA CLNPASPMVK KIIEKMLKNG KSN
  • Endotoxin level:
    Low
  • Formulation:
    0,1% Trifluoroacetic Acid (TFA)
  • Nuclease-free:
    N
  • Grade:
    RUO
  • Shipping temperature:
    RT

Specifications

Frequently Bought Together