Order Entry
Switzerland
ContactUsLinkComponent
Indeno[1,2,3-cd]pyrene
Indeno[1,2,3-cd]pyrene
Catalog # CAMBULM-2426-25
CAS Number:  
Indeno[1,2,3-cd]pyrene
Catalog # CAMBULM-2426-25
CAS Number:  

Specifications

  • Pack type:
    Glass bottle for solids
  • Pk:
    25 mg
  • Seq:
    2315

Specifications

About this item

 :
 :276.34 g/mol
Boiling Pt:536 °C (1013 hPa)
Melting Pt:163…164 °C
EINECS:205-893-2
 :GHS07
CAS Number:193-39-5
Formula:C₂₂H₁₂