

1-Methylimidazole (NMI) ≥99.0% (by GC) for synthesis
: Merck
:616-47-7
:
:C₄H₆N₂
:MFCD00005292
:198 °C (1013 hPa)
:–2 °C
:1.03 g/cm³ (20 °C)
:92 °C
:210-484-7
:2922
:8 (6.1),II
:M-Clarity™ quality level = MQ200
:82.11 g/mol
:GHS06
:GHS05
:Ambient
- Solubility:(20 °C) soluble
- Melting Point:-2 °C
- Molar Mass:82.10 g/mol
- Boiling Point:198 °C (1013 hPa)
- Vapor Pressure:0.4 hPa (20 °C)
- Flash Point:92 °C
- Explosion Limit:2.7 - 15.7 %(V)
- Density:1.035 g/cm3 (20 °C)
- pH:9.5 - 10.5 (50 g/l, H2O, 20 °C)
- Ignition Point:525 °C
Frequently Bought Together