


Imidazole ≥99%, GPR RECTAPUR®
Catalog # 24720.157
Supplier: VWR Chemicals
CAS Number:
Specifications
- Pack type:Plastic bottle for solids
- Pk:50 g
- Seq:3368
Specifications
Specification Test Results
- Assay:Min. 99 %
- Melting point:88 - 91 °C
- pH (20°C; 5 %):9.5 - 10.5
- Ignition residue (SO4):Max. 0.1 %
- Water:Max. 0.5 %
- Fe (Iron):Max. 10 ppm
Specification Test Results
About this item
:
Merck Index:13,04935
Boiling Pt:255 °C (1013 hPa)
Melting Pt:89…90 °C
Density:1.18 g/cm³ (20 °C)
Flash Pt:135 °C
Storage Temperature:Ambient
EINECS:206-019-2
UN:2923
ADR:8 (6.1),III
:68.08 g/mol
:GHS08
:GHS07
:GHS05
CAS Number:288-32-4
Formula:C₃H₄N₂
MDL Number:MFCD00005183