


Imidazole for synthesis, Sigma-Aldrich®
Supplier: Merck
Certificates
About this item
CAS Number:288-32-4
:
Formula:C₃H₄N₂
MDL Number:MFCD00005183
Merck Index:13,04935
Boiling Pt:255 °C (1013 hPa)
Melting Pt:89…90 °C
Density:1.18 g/cm³ (20 °C)
Flash Pt:135 °C
EINECS:206-019-2
UN:3263
ADR:8,III
:M-Clarity™ quality level = MQ200
:68.08 g/mol
:GHS05
:GHS08
:GHS07
Storage Temperature:Ambient
Specification Test Results
- Solubility:633 g/l (20 °C)
- Melting Point:89 - 90 °C
- Molar Mass:68.08 g/mol
- Bulk Density:500 - 600 kg/m3
- Boiling Point:256 °C (1013 hPa)
- Vapor Pressure:0.003 hPa (20 °C)
- Flash Point:145 °C
- Density:1.030 g/cm3
- pH:10.5 (67 g/l, H2O, 20 °C)
- Ignition Point:480 °C
- Hazard Class:8
- Hazard Class Description:Corrosive Material
Specification Test Results
Related Information
Frequently Bought Together