Order Entry
Switzerland
ContactUsLinkComponent
Imidazole, BAKER ANALYZED®, Laboratory Reagent, J.T.Baker®
Imidazole, BAKER ANALYZED®, Laboratory Reagent, J.T.Baker®
Supplier:  Avantor
Certificates
undefined
Imidazole, BAKER ANALYZED®, Laboratory Reagent, J.T.Baker®
Supplier:  Avantor
Certificates

Specification Test Results

  • Assay:
    min. 99.5 %
  • Iron (Fe):
    max. 0.001 %
  • Heavy Metals (as Pb):
    max. 0.001 %
  • Melting Point:
    88 - 90 °C
  • Residue after Ignition:
    max. 0.1 %

Specification Test Results

Related Information

Frequently Bought Together