

Iodine trichloride (Wijs'reagent) for determination of iodine value, traceable to NIST Standard Reference Material (SRM)
Supplier: VWR Chemicals
Certificates
About this item
CAS Number:865-44-1
:
Formula:ICl₃
MDL Number:MFCD00036291
:233.26 g/mol
Melting Pt:63 °C
Density:1.23 g/cm³ (20 °C)
Flash Pt:40 °C
Storage Temperature:Ambient
EINECS:212-739-8
UN:3085
ADR:5.1 (8),II
:GHS05
:GHS07
Specification Test Results
- Concentration:0.0994 - 0.1006 mol/l
- Form:Liquid
- NIST traceable:Confirmed
Specification Test Results
Related Information
Certificates
Search by Lot Number1L Glass bottle for liquids DIN 45 (ZPACK13415101) 500 ml Glass bottle for liquids DIN 45 (ZPACE8009063)
Packaging Information
spec_VWRC2812.0500 spec_VWRC2812.1000
Product specification
Frequently Bought Together