
Diphenyl sulphide ≥98%
Supplier: Thermo Fisher Scientific
Certificates
About this item
CAS Number:139-66-2
:
Formula:C₁₂H₁₀S
MDL Number:MFCD00003064
:186.28 g/mol
Merck Index:14,7314
Boiling Pt:295…296 °C
Melting Pt:–40 °C
Density:1.118
Flash Pt:>110 °C (230 °F)
Storage Temperature:Ambient
EINECS:205-371-4
UN:2810
ADR:6.1,III
:GHS07
Related Information
Frequently Bought Together