
Dibutylsulfid ≥98%
Supplier: Thermo Fisher Scientific
Certificates
About this item
CAS Number:544-40-1
:
Formula:C₈H₁₈S
MDL Number:MFCD00009468
:146.3 g/mol
Merck Index:14,1590
Boiling Pt:188…189 °C
Melting Pt:–80 °C
Density:0.839
Flash Pt:76 °C (168 °F)
Storage Temperature:Raumtemperatur
EINECS:208-870-5
UN:3334
ADR:9,III
:GHS07
Related Information
Frequently Bought Together