Tubing, for milk and dairy dispensing, Tygon S3™ B-44-4X
Tygon S3™ B-44-4X is a phthalate-free flexible tubing for food and beverage dispensing.
- Smooth, non-porous bore will not trap particulates or promote bacterial growth
- Resistant to harsh alkaline cleaners and sanitizers
- Excellent alternative to rigid piping systems
Temperature resistance: −36 to +74 °C
Hardness (Shore durometer A): 66
Density: 1,21
Frequently Bought Together