Dimethyl sulphoxide, Uvasol® for spectroscopy, Supelco®
Supplier: Merck
Certificates
About this item
CAS Number:67-68-5
:
Formula:(CH₃)₂SO
MDL Number:MFCD00002089
:78.14 g/mol
Merck Index:13,03285
Boiling Pt:189 °C (1013 hPa)
Melting Pt:18.5 °C
Density:1.101 g/cm³ (20 °C)
:M-Clarity™ quality level = MQ100
Flash Pt:87 °C
Storage Temperature:Ambient
EINECS:200-664-3
Specifications
- Environmentally Preferable:Safer Human and Environmental Health
- Environmental certification:Chem21 Solvent Selection Guide recommended by default
Specifications
Specification Test Results
- Solubility:1000 g/L (20 °C)
- Melting Point:18.4 °C
- Molar Mass:78.1 g/mol
- Boiling Point:189 °C (1013 hPa)
- Vapour Pressure:0.6 hPa (20 °C)
- Flash Point:87 °C
- Refractive Index:1.48 (20 °C)
- Explosion Limit:1.8 - 63.0%(V)
- Density:1.10 g/cm³ (20 °C)
- Ignition Point:301 °C
- Saturation Concentration:8.0 g/m³ (20 °C)
- Viscosity Kinematic:2.14 mm²/s
Specification Test Results
Related Information
Certificates
Search by Lot NumberMERC1.02950.2500_TDS_EN ProductPage_102950
Produktspezifikation
msds_102950
Sicherheitsdatenblatt (SDS)
Solvents for spectroscopy Uvasol
Produktliteratur
Frequently Bought Together