ContactUsLinkComponent
Dimethyl sulphoxide 99.8+% for peptide synthesis
Dimethyl sulphoxide 99.8+% for peptide synthesis
Certificates
undefined
Dimethyl sulphoxide 99.8+% for peptide synthesis
Specification Test Results
  • Appearance:
    Clear colorless Liquid
  • Water:
    ≤0.005 % (K.F.)
  • GC:
    ≥99.80 %
  • Residue after evaporation:
    ≤0.001 %
  • Free amine:
    ≤0.0002 %
  • Acidity (CH3COOH):
    ≤0.003 %