Order Entry
Switzerland
ContactUsLinkComponent
Selenium disulphide for synthesis, Sigma-Aldrich®
Toxic
Health
Environment
Selenium disulphide for synthesis, Sigma-Aldrich®
Catalog # 8.43845.0025
Supplier:  Merck
CAS Number:  
Toxic
Health
Environment
Selenium disulphide for synthesis, Sigma-Aldrich®
Catalog # 8.43845.0025
Supplier:  Merck
CAS Number:  

Specifications

  • Pack type:
    Glass bottle
  • Pk:
    25 g
  • Seq:
    3308

Specifications

Specification Test Results

  • Solubility:
    (20 °C) insoluble
  • Molar Mass:
    143.09 g/mol
  • Hill Formula:
    S2Se
  • Chemical Formula:
    SeS2

Specification Test Results

About this item

 :
 :143.09 g/mol
Boiling Pt:–112 °C (1013 hPa)
Density:0.03 g/cm³ (–65 °C)
ADR:6.1,II
 :M-Clarity™ quality level = MQ200
EINECS:231-303-8
UN:3283
 :GHS06
 :GHS08
 :GHS09
Storage Temperature:Ambient
CAS Number:7488-56-4
Formula:S₂Se
MDL Number:MFCD00011227