
Ammonium sulphamate, EMSURE® ACS, Reag. Ph. Eur. for analysis, Supelco®
Catalog # 1.01220.0100
Supplier: Merck
CAS Number:
Specifications
- Pack type:Plastic bottle
- Pk:100 g
- Seq:3401
Specifications
Specification Test Results
- Solubility:1950 g/l (20 °C)
- Melting Point:132 - 134 °C
- Molar Mass:114.13 g/mol
- Bulk Density:680 kg/m3
- Boiling Point:160 °C (1013 hPa) decomposes
- pH:5.0 - 6.0 (50 g/l, H2O, 20 °C)
Specification Test Results
About this item
:
:114.13 g/mol
Merck Index:12,00589
Melting Pt:131.1 °C
Density:>1 g/cm³ (20 °C)
:M-Clarity™ quality level = MQ300
EINECS:231-871-7
:GHS07
CAS Number:7773-06-0
Formula:H₆N₂O₃S
MDL Number:MFCD00011429