Diammonium hydrogen orthophosphate ≥97.5%, TECHNICAL
Catalog # 21306.362
Supplier: VWR Chemicals
CAS Number:
Specifications
- Pack type:Plastic bucket
- Pk:5 kg
- Seq:4114
- Environmentally Preferable:
Specifications
Specification Test Results
- Assay:Min. 97.5 %
Specification Test Results
About this item
Synonyms: Diammonium hydrogen orthophosphate, Ammonium hydrogen phosphate, di-Ammonium hydrogen phosphate, Ammonium Phosphate, Ammonium biphosphate, Fyrex, Ammonium phosphate dibasic, Ammonium monohydrogen phosphate, Diammonium hydrogen phosphate
:
:132.06 g/mol
Merck Index:13,00546
Melting Pt:185 °C
Density:1.619 g/cm³ (20 °C)
Storage Temperature:Ambient
EINECS:231-987-8
REACH:01-2119490974-22
CAS Number:7783-28-0
Formula:(NH₄)₂HPO₄
MDL Number:MFCD00010891