
Ammonium chloride ≥99%, GPR RECTAPUR®
Supplier: VWR Chemicals
Certificates
About this item
CAS Number:12125-02-9
:
Formula:NH₄Cl
MDL Number:MFCD00011420
:53.49 g/mol
Merck Index:13,00510
Melting Pt:335 °C
Density:1.5256 g/cm³ (20 °C)
EINECS:235-186-4
REACH:01-2119487950-27
:GHS07
Storage Temperature:Ambient
Specification Test Results
- Assay:Min. 99 %
- Heavy metals (as Pb):Max. 10 ppm
- Ignition residue (SO4):Max. 0.9 %
- SO4 (Sulphate):Max. 100 ppm
- Fe (Iron):Max. 10 ppm
Specification Test Results
Related Information
Certificates
Search by Lot Number2,5L Plastic bottle for solids DIN 80 (ZPACK13252500) 33L Plastic bucket UN Z (ZPACK13247037)
Packaging Information
21235_AMMONIUM CHLORIDE_BSE_REV0.pdf
BSE/TSE Certificates
SDS_EN_CH_VWRC21235.pdf SDS_EN_DE_VWRC21235.pdf SDS_EN_NL_VWRC21235.pdf SDS_EN_UK_VWRC21235.pdf sds_VWRC21235
SDS
spec_VWRC21235.297 spec_VWRC21235.366 spec_VWRC21235.366 spec_VWRC21235.468 spec_VWRC21235.468
Product specification
VWR Chemicals Big Packs and Bulks
Product Literature
Frequently Bought Together