Order Entry
Switzerland
ContactUsLinkComponent
Triisopropylsilane for synthesis, Sigma-Aldrich®
Flammable
Triisopropylsilane for synthesis, Sigma-Aldrich®
Catalog # 8.41359.0010
Supplier:  Merck
CAS Number:  
Flammable
Triisopropylsilane for synthesis, Sigma-Aldrich®
Catalog # 8.41359.0010
Supplier:  Merck
CAS Number:  

Specifications

  • Pack type:
    Glass bottle
  • Pk:
    10 ml
  • Seq:
    2275

Specifications

Specification Test Results

  • Molar Mass:
    158.35 g/mol
  • Boiling Point:
    84 - 85 °C (46.7 hPa)
  • Flash Point:
    35 °C
  • Density:
    0.77 g/cm3 (20 °C)

Specification Test Results

About this item

 :
 :158.36 g/mol
Boiling Pt:84 °C (1013 hPa)
Density:0.7734 g/cm³ (20 °C)
Storage Temperature:Ambient
ADR:3,II
 :M-Clarity™ quality level = MQ200
Flash Pt:35 °C
UN:1993
 :GHS02
CAS Number:6485-79-6
Formula:C₉H₂₂Si
MDL Number:MFCD00009657