


Carbon disulphide ≥99.9% (by GC) ACS, Riedel-de Haën™
:75-15-0
:
:CS₂
:MFCD00011321
:76.14 g/mol
:12,01859
:46 °C (1.013 hPa)
:–76 °C
:1.264 g/cm³ (20 °C)
:–30 °C
:Ambient
:200-843-6
:1131
:3,I
:GHS02
:GHS08
:GHS07
- Identity (IR):complying
- Assay (GC):Min. 99.90 %
- Non-volatile matter:Max. 0.002 %
- Water (Karl Fischer):Max. 0.05 %
- Sulfur dioxide (SO₂):complying
- Hydrogen sulfide (H₂S):complying
- APHA:Max. 10
- Autoignition Temperature:212 °F
- Refractive Index:n20/D 1.627 (lit.)
- Explosion Limit:50 %
Frequently Bought Together