Tetraethyl orthosilicate (TEOS) ≥99%, GPR RECTAPUR®
Catalog # 24004.290
Supplier: VWR Chemicals
CAS Number:
Specifications
- Pack type:Glass bottle
- Pk:1 L
- Seq:5037
Specifications
Specification Test Results
- Assay:Min. 99 %
- Acidity:Max. 0.002 meq/g
- Density (20/4):0.930 - 0.940
- Cl (Chloride):Max. 0.02 %
Specification Test Results
About this item
:
:208.33 g/mol
Merck Index:13,03882
Boiling Pt:168 °C (1013 hPa)
Melting Pt:–85,5 °C
Density:0.933 g/cm³ (20 °C)
Flash Pt:38 °C
Storage Temperature:Ambient
EINECS:201-083-8
UN:1292
ADR:3,III
:GHS02
:GHS07
CAS Number:78-10-4
Formula:Si(OC₂H₅)₄
MDL Number:MFCD00009062