Lead(IV) oxide
Glass bottle for solids, 100 g
Catalog # ROTH4479.1
Marketsource
Supplier: Roth Carl
Jump to:
Product Details & Documents
Synonyms: Lead dioxide
- CAS Number:1309-60-0
- Formula:O₂Pb
- MDL Number:MFCD00011165
- Molecular Weight:239.2 g/mol
- Merck Index:12,05424
- Melting Pt:290 °C
- Density:9.4 g/cm³ (20 °C)
- EINECS:215-174-5
- UN:1872
- ADR:5.1,III
Specifications
Specifications
- Pack type:Glass bottle for solids
- Pk:100 g



