Order Entry
Switzerland
ContactUsLinkComponent
Carborundum, TECHNICAL 0.105 mm
Carborundum, TECHNICAL 0.105 mm
Catalog # 22505.297
Supplier:  VWR Chemicals
CAS Number:  
undefined
Carborundum, TECHNICAL 0.105 mm
Catalog # 22505.297
Supplier:  VWR Chemicals
CAS Number:  

Specifications

  • Pack type:
    Plastic bottle for solids
  • Pk:
    1 kg
  • Seq:
    4024

Specifications

Specification Test Results

  • Identification:
    Passes test

Specification Test Results

About this item

 :
 :40.1 g/mol
Merck Index:12,08636
Boiling Pt:< 2700 °C (1013 hPa)
Melting Pt:2700 °C
Density:3.23 g/cm³ (20 °C)
EINECS:206-991-8
CAS Number:409-21-2
Formula:SiC