Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
0
SFr. 0.00
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Switzerland
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Home
Carborundum
Carborundum, TECHNICAL 1.190 mm
Carborundum, TECHNICAL 1.190 mm
22490.295
:
VWR Chemicals
:
Carborundum, TECHNICAL 1.190 mm
22490.295
:
VWR Chemicals
:
Pack type:
Plastic bottle for solids
Pk:
1 kg
Seq:
4024
Identification:
Passes test
:
:
40.1 g/mol
:
12,08636
:
< 2700 °C (1013 hPa)
:
2700 °C
:
3.23 g/cm³ (20 °C)
:
206-991-8
:
409-21-2
:
SiC