Carborundum, TECHNICAL 1.680 mm
Catalog # 22488.297
Supplier: VWR Chemicals
CAS Number:
Specifications
- Pack type:Plastic bottle for solids
- Pk:1 kg
- Seq:4024
Specifications
Specification Test Results
- Identification:Passes test
Specification Test Results
About this item
:
:40.1 g/mol
Merck Index:12,08636
Boiling Pt:< 2700 °C (1013 hPa)
Melting Pt:2700 °C
Density:3.23 g/cm³ (20 °C)
EINECS:206-991-8
CAS Number:409-21-2
Formula:SiC