Order Entry
Switzerland
ContactUsLinkComponent
Aluminium hydroxide, powder, extra pure
Irritant
Aluminium hydroxide, powder, extra pure
Certificats
undefined
Irritant
Aluminium hydroxide, powder, extra pure

Résultats des tests de spécification

  • Appearance:
    White to almost white Powder
  • Assay:
    ≥63.5 % (Al2O3)
  • Soluble matter:
    ≤0.2 % (in water)
  • Loss on ignition:
    ≤36.5 %

Résultats des tests de spécification

Informations associées

Frequently Bought Together