Order Entry
Switzerland
ContactUsLinkComponent
Magnesium sulphate hydrate, dried 99.0-101.0% (dried basis), Puriss., meets analytical specification of DAC, Fluka™
Magnesium sulphate hydrate, dried 99.0-101.0% (dried basis), Puriss., meets analytical specification of DAC, Fluka™
Certificates
Magnesium sulphate hydrate, dried 99.0-101.0% (dried basis), Puriss., meets analytical specification of DAC, Fluka™

Specification Test Results

  • Assay (calculated to the dried substance):
    99.0 - 101.0 %
  • Loss on drying (400°C):
    26.0 - 32.0 %
  • Arsenic (As):
    Max. 2 ppm
  • Iron (Fe):
    Max. 10 ppm
  • Heavy metals (as Pb):
    Max. 15 ppm
  • Chloride (Cl):
    Max. 400 ppm
  • Acidity or alkalinity:
    complying
  • Appearance of the solution:
    complying
  • Residual solvents:
    complying

Specification Test Results

Related Information

Frequently Bought Together