Magnesium oxide, 2 part ceramic adhesive paste
Anbieter: Thermo Fisher Scientific
Zertifikate
Über diesen Artikel
High temperature adhesive for bonding ceramic to metal.
High temperature adhesive for bonding ceramic to metal.
Mix ratio (w/w): 1.5:1 powder to liquid.
Cure conditions: 1-4 hours at room temperature, 2 hours at 94 °C
High temperature adhesive for bonding ceramic to metal
Frequently Bought Together