SFr. 0.00
Order Entry
/
Products
Services
Digital Solutions
Applications
Support
About
Switzerland
ContactUsLinkComponent
Magnesium oxide
Magnesium oxide substrate 10x10x0.5mm, polished one side 110 orientation
Magnesium oxide substrate 10x10x0.5mm, polished one side 110 orientation
40272.KU
:
Thermo Fisher Scientific
:
Magnesium oxide substrate 10x10x0.5mm, polished one side 110 orientation
40272.KU
:
Thermo Fisher Scientific
:
Pk:
5
Seq:
0005
:
:
40.3 g/mol
:
13,05700
:
3600 °C (1013 hPa)
:
2800 °C
:
3.58 g/cm³ (25 °C)
:
Ambient
:
215-171-9
:
1309-48-4
:
MgO
:
MFCD00011109