Calciumfluorid, wasserfrei 99%, rein
Supplier: Thermo Fisher Scientific
Certificates
About this item
CAS Number:7789-75-5
:
Formula:CaF₂
MDL Number:MFCD00010907
:78.07 g/mol
Merck Index:13,01669
Boiling Pt:2500 °C (760 mmHg)
Melting Pt:1402 °C
Density:3.18 g/cm³
EINECS:232-188-7
:GHS07
Storage Temperature:Raumtemperatur
Specification Test Results
- Appearance:White to off-white Powder
- Assay:≥98.5 % (CaF2)
Specification Test Results
Frequently Bought Together