Order Entry
Switzerland
ContactUsLinkComponent
Ammonium hydrogen difluoride ≥97%, TECHNICAL
Toxic
Corrosive
Ammonium hydrogen difluoride ≥97%, TECHNICAL
Supplier:  VWR Chemicals
Certificates
undefined
Toxic
Corrosive
Ammonium hydrogen difluoride ≥97%, TECHNICAL
Supplier:  VWR Chemicals
Certificates

Specification Test Results

  • Assay:
    Min. 97 %

Specification Test Results

Frequently Bought Together