Order Entry
Switzerland
ContactUsLinkComponent
 
 
 
 
Potassium peroxodisulphate ≥99.0% ACS, Fluka™
 
 
 
 
Potassium peroxodisulphate ≥99.0% ACS, Fluka™

 

  • Assay:
    99.0 - 101.0 %
  • Water insoluble matter:
    Max. 0.005 %
  • Iron (Fe):
    Max. 5 ppm
  • Manganese (Mn):
    Max. 1 ppm
  • Heavy metals (as Pb):
    Max. 0.001 %
  • Chlorine compounds (Cl):
    Max. 10 ppm
  • Vapor Density:
    9.3 (vs air)

 

 

Frequently Bought Together