


Potassium peroxodisulphate, EMSURE® ACS, Reag. Ph. Eur. for analysis, Supelco®
: Merck
:7727-21-1
:
:K₂S₂O₈
:MFCD00011386
:270.32 g/mol
:13,07741
:100 °C
:2.477 g/cm³ (20 °C)
:Ambient
:231-781-8
:5.1,III
:M-Clarity™ quality level = MQ300
:1492
:GHS03
:GHS08
:GHS07
- Solubility:50 g/l (20 °C)
- Melting Point:100 °C (decomposition)
- Molar Mass:270.32 g/mol
- Bulk Density:1150 kg/m3
- Density:2.48 g/cm3 (20 °C)
- pH:4 - 5 (50 g/l, H2O, 20 °C)
- Hazard Class:5.1
- Hazard Class Description:Oxidizer
Frequently Bought Together