


Potassium peroxodisulphate ≥99.0% ACS
Supplier: Thermo Fisher Scientific
Certificates
About this item
CAS Number:7727-21-1
:
Formula:K₂S₂O₈
MDL Number:MFCD00011386
:270.32 g/mol
Merck Index:14,7656
Melting Pt:100 °C (dec.)
Density:2.477
EINECS:231-781-8
UN:1492
ADR:5.1,III
:GHS08
:GHS03
:GHS07
Storage Temperature:Ambient
Related Information
Frequently Bought Together