Kaliumdisulfat, EMSURE® ACS zur Analyse, Supelco®
Supplier: Merck
Certificates
About this item
CAS Number:7790-62-7
:
Formula:K₂O₇S₂
MDL Number:MFCD00011385
:254.32 g/mol
Merck Index:13,07749
Melting Pt:∼325 °C
Density:2.51 g/cm³ (20 °C)
Storage Temperature:Raumtemperatur
EINECS:232-216-8
ADR:8 (6.1),I
:M-Clarity™ quality level = MQ300
UN:2923
:GHS06
:GHS05
Specification Test Results
- Assay (acidimetric, as K₂S₂O₇):97.5 - 100.0 %
- Acidity (as H₂SO₄):37.5 - 38.6 %
- Insoluble and by ammoniumhydroxide precipitated matter:≤ 0.01 %
- Chloride (Cl):≤ 0.0005 %
- Phosphate (PO₄):≤ 0.001 %
- Total nitrogen (N):≤ 0.002 %
- Heavy metals (as Pb):≤ 0.001 %
- Al (Aluminium):≤ 0.001 %
- As (Arsenic):≤ 0.0005 %
- Ca (Calcium):≤ 0.002 %
- Fe (Iron):≤ 0.0005 %
- Mg (Magnesium):≤ 0.0005 %
- Na (Sodium):≤ 0.01 %
- Water (H₂O):≤ 2.5 %
Specification Test Results
Related Information
Frequently Bought Together