Order Entry
Switzerland
ContactUsLinkComponent
Sodium dihydrogen phosphate ≥99.0% for HPLC, Fluka™
Sodium dihydrogen phosphate ≥99.0% for HPLC, Fluka™
Certificates
Sodium dihydrogen phosphate ≥99.0% for HPLC, Fluka™
Certificates

Specification Test Results

  • Assay:
    99.0 - 101.0 %
  • Appearance of the solution (c=10%):
    complying
  • pH (10 %, 20°C):
    4.0 - 4.5
  • Absorbance at 210 nm:
    Max. 0.1
  • Absorbance at 260 nm:
    Max. 0.06
  • Absorbance 280 nm:
    Max. 0.04
  • Absorbance at 320 nm:
    Max. 0.02
  • Absorbance 500 nm:
    Max. 0.02

Specification Test Results

Related Information

Frequently Bought Together