Order Entry
Switzerland
ContactUsLinkComponent
Potassium dihydrogen phosphate ≥99.5%, HiPerSolv CHROMANORM® for HPLC
Potassium dihydrogen phosphate ≥99.5%, HiPerSolv CHROMANORM® for HPLC
  153184U
 :  
undefined
Potassium dihydrogen phosphate ≥99.5%, HiPerSolv CHROMANORM® for HPLC
  153184U
 :  

 

  • Pack type:
    Plastic bottle for solids
  • Pk:
    500 g
  • Seq:
    3645

 

 

  • Assay:
    Min. 99.5 %
  • pH (20°C; 5 %):
    4.3 - 4.5
  • Heavy metals (as Pb):
    Max. 10 ppm
  • Cl (Chloride):
    Max. 5 ppm
  • SO4 (Sulphate):
    Max. 30 ppm
  • Fe (Iron):
    Max. 10 ppm
  • Transmittance (254 nm) (0.1 mol/l):
    Min. 90 %

 

 

 :
 :136.09 g/mol
 :13,07744
 :~100 °C (1013 hPa)
 :253 °C
 :2.3 g/cm³ (20 °C)
 :Ambient
 :231-913-4
 :01-2119490224-41
 :7778-77-0
 :KH₂PO₄
 :MFCD00011401