Order Entry
Switzerland
Orders LinkContactUsLinkComponent
Potassium nitrate

Potassium nitrate

Oxidizing
Marketsource
Supplier:   Roth Carl
Potassium nitrate
Supplier:  Roth Carl
Marketsource
Certificates
Potassium nitrate

3 Options Available Below

Product Details & Documents
Synonyms:  Nitrate of Potash
  • CAS Number:7757-79-1
  • Formula:KNO₃
  • MDL Number:MFCD00011409
  • Molecular Weight:101.1 g/mol
  • Merck Index:13,07733
  • Melting Pt:334 °C
  • Density:2.109 g/cm³ (20 °C)
  • Storage Temperature:Ambient
  • EINECS:231-818-8
  • UN:1486
  • ADR:5.1,III
  • REACH:01-2119488224-35

Documents

Product Family Options

Product Information

  • Pack type
    Pk
    Availability
    Price
  • Glass bottle for solids
    500 g
    Log in for availability
    Log in for availability
    1 * 500 g
    1 * 500 g - SFr. 22.20
    SFr. 22.20
    1 * 500 g
       SFr. 22.20

    Specifications:
    Marketsource
    Marketsource
  • Glass bottle for solids
    1 kg
    Log in for availability
    Log in for availability
    1 * 1 kg
    1 * 1 kg - SFr. 33.00
    SFr. 33.00
    1 * 1 kg
       SFr. 33.00

    Specifications:
    Marketsource
    Marketsource
  • Glass bottle for solids
    2,5 kg
    Log in for availability
    Log in for availability
    1 * 2,5 kg
    1 * 2,5 kg - SFr. 63.00
    SFr. 63.00
    1 * 2,5 kg
       SFr. 63.00

    Specifications:
    Marketsource
    Marketsource
Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.