Order Entry
Switzerland
Orders LinkContactUsLinkComponent
 

Potassium nitrate

Oxidizing
Supplier:   EUROCLONE
Potassium nitrate
Supplier:  EUROCLONE
Certificates
Potassium nitrate

3 Options Available Below

Product Details & Documents
Synonyms:  Nitrate of Potash
  • CAS Number:7757-79-1
  • Formula:KNO₃
  • MDL Number:MFCD00011409
  • Molecular Weight:101.1 g/mol
  • Merck Index:13,07733
  • Melting Pt:334 °C
  • Density:2.109 g/cm³ (20 °C)
  • Storage Temperature:Ambient
  • EINECS:231-818-8
  • UN:1486
  • ADR:5.1,III
  • REACH:01-2119488224-35
Product Family Options

Product Information

  • Pack type
    Pk
    Availability
    Price
  • 1 KIT
    Log in for availability
    Log in for availability
    1 * 1 KIT
    1 * 1 KIT

  • Glass bottle for solids
    1 kg
    Log in for availability
    Log in for availability
    1 * 1 kg
    1 * 1 kg

  • 5 kg
    Log in for availability
    Log in for availability
    1 * 5 kg
    1 * 5 kg