

Kaliumperchlorat, EMSURE® ACS zur Analyse, Supelco®
Supplier: Merck
Certificates
About this item
CAS Number:7778-74-7
:
Formula:ClKO₄
MDL Number:MFCD00011362
:138.55 g/mol
Merck Index:13,07738
Melting Pt:608 °C
Density:2.5239 g/cm³ (10.7 °C)
EINECS:231-912-9
ADR:5.1,II
:M-Clarity™ quality level = MQ300
UN:1489
:GHS03
:GHS07
Specification Test Results
- Solubility:17 g/l (20 °C)
- Melting Point:610 °C (decomposition)
- Molar Mass:138.55 g/mol
- Bulk Density:1150 kg/m3
- pH:5.0 (10 g/l, H2O, 25 °C)
- Hazard Class:5.1
- Hazard Class Description:Oxidizer
Specification Test Results
Related Information
Frequently Bought Together