Lithium chloride
Marketsource
Supplier: Roth Carl
Jump to:
About this item
- CAS Number:7447-41-8
- Formula:LiCl
- MDL Number:MFCD00011078
- Molecular Weight:42.39 g/mol
- Merck Index:13,05550
- Boiling Pt:1382 °C (1013 hPa)
- Melting Pt:614 °C
- Density:2.07 g/cm³ (20 °C)
- Storage Temperature:Ambient
- EINECS:231-212-3
2 Options Available Below
Product Details & Documents
- CAS Number:7447-41-8
- Formula:LiCl
- MDL Number:MFCD00011078
- Molecular Weight:42.39 g/mol
- Merck Index:13,05550
- Boiling Pt:1382 °C (1013 hPa)
- Melting Pt:614 °C
- Density:2.07 g/cm³ (20 °C)
- Storage Temperature:Ambient
- EINECS:231-212-3



