Order Entry
Switzerland
ContactUsLinkComponent
 
Sodium bromide ≥99.0% ACS, Fluka™
  310506-500G
 :  
Sodium bromide ≥99.0% ACS, Fluka™
  310506-500G
 :  

 

  • Pack type:
    Plastic bottle
  • Pk:
    500 g
  • Seq:
    3645

 

 

  • Assay:
    Min. 99.0 %
  • pH (5 %, 25 °C):
    5.0 - 8.8
  • Water insoluble matter:
    Max. 0.005 %
  • Barium (Ba):
    Max. 0.002 %
  • Calcium (Ca):
    Max. 0.002 %
  • Iron (Fe):
    Max. 5 ppm
  • Potassium (K):
    Max. 0.1 %
  • Magnesium (Mg):
    Max. 0.001 %
  • Heavy metals (as Pb):
    Max. 5 ppm
  • Bromate (BrO₃):
    Max. 0.001 %
  • Chloride (Cl):
    Max. 0.2 %
  • Sulfate (SO₄):
    Max. 0.002 %

 

 

 :
 :102.89 g/mol
 :13,08666
 :1.393 °C (1.013 hPa)
 :750 °C
 :3.2 g/cm³ (25 °C)
 :Ambient
 :231-599-9
 :7647-15-6
 :NaBr
 :MFCD00003475