Order Entry
Switzerland
ContactUsLinkComponent
 
 
Sulphuric acid 95.0-98.0% ACS, Fluka™
  320501-1L
 :  
 
Sulphuric acid 95.0-98.0% ACS, Fluka™
  320501-1L
 :  

 

  • Pack type:
    Glass bottle
  • Pk:
    1 L
  • Seq:
    5037

 

 

  • Assay:
    95.0 - 98.0 %
  • Density (D 20/4):
    1.825 - 1.836
  • Residue on ignition:
    Max. 5 ppm
  • Ammonium (NH₄):
    Max. 2 ppm
  • Arsenic (As):
    Max. 0.01 ppm
  • Iron (Fe):
    Max. 0.2 ppm
  • Mercury (Hg):
    Max. 0.005 ppm
  • Heavy metals (as Pb):
    Max. 1 ppm
  • Chloride (Cl):
    Max. 0.2 ppm
  • Nitrate (NO₃):
    Max. 0.5 ppm
  • KMnO₄ reducing matter (as SO₂):
    Max. 2 ppm
  • APHA:
    Max. 10
  • Appearance:
    complying

 

 

 :
 :98.08 g/mol
 :14,08974
 :310 °C (1.013 hPa)
 :–14…–10 °C
 :1.840 g/cm³ (20 °C)
 :Ambient
 :231-639-5
 :1830
 :8,II
 :GHS05
 :7664-93-9
 :H₂SO₄
 :MFCD00064589