Order Entry
Switzerland
ContactUsLinkComponent
Fluorescein free acid Reag. Ph. Eur., Fluka™
Fluorescein free acid Reag. Ph. Eur., Fluka™
Catalog # 32615-100G
CAS Number:  
undefined
Fluorescein free acid Reag. Ph. Eur., Fluka™
Catalog # 32615-100G
CAS Number:  

Specifications

  • Pack type:
    Glass bottle
  • Pk:
    100 g
  • Seq:
    3401

Specifications

Specification Test Results

  • Ignition Residue:
    ≤0.25% (as SO₄)
  • Identity (IR):
    complying
  • UV spectrum:
    complying

Specification Test Results

About this item

 :
 :332.31 g/mol
Merck Index:13,04185
Melting Pt:314-316 °C
Melting Pt:314…316 °C
Storage Temperature:Ambient
EINECS:219-031-8
CAS Number:2321-07-5
Formula:C₂₀H₁₂O₅
MDL Number:MFCD00005050