Fluorescein
Catalog # MOLE36797905
Supplier: Molekula
CAS Number:
Specifications
- Pack type:Glass bottle for solids
- Pk:500 g
- Seq:3645
Specifications
About this item
:
:332.31 g/mol
Merck Index:13,04185
Melting Pt:314…316 °C
Storage Temperature:Ambient
EINECS:219-031-8
CAS Number:2321-07-5
Formula:C₂₀H₁₂O₅
MDL Number:MFCD00005050