Fluorescein 99%, pure, laser grade
Catalog # ACRO410620010
Supplier: Thermo Fisher Scientific
CAS Number:
Specifications
- Pack type:Glass bottle for solids
- Pk:1 g
- Seq:3047
Specifications
Specification Test Results
- Appearance:Orange to red to brown Powder
- Infrared spectrum:Conforms
- HPLC:≥98.5 %
- UV Lambda max:496 to 500 nm (in Basic Methanol)
- Suitability:(for use as a laser dye) Passes
- Molar absorption coefficient:≥63900 l/(mol.cm) (at Lambda max)
Specification Test Results
About this item
:
:332.31 g/mol
Merck Index:13,04185
Melting Pt:314…316 °C
EINECS:219-031-8
:GHS07
Storage Temperature:Ambient
CAS Number:2321-07-5
Formula:C₂₀H₁₂O₅
MDL Number:MFCD00005050