Dantron, Neat
Catalog # PROOBP428
Supplier: LGC Standards PROMOCHEM
CAS Number:
Specifications
- Pack type:Glass bottle
- Pk:50 mg
- CAS No.:117-10-2
- Description:Dantron, Neat
- Synonym(s):1,8-Dihydroxyanthracene-9,10-dione, Dithranol Imp. B (EP), Danthron, 1,8-Dihydroxy-9,10-anthraquinone, 1,8-Dihydroxyanthraquinone, 1,8-Dioxyanthraquinone, Altan, Antrapurol, Chrysazin, Danthrone, Dantron, Diaquone, Dionone, Dorbane, Istin, Istizin, Laxanorm, Laxanthreen, Laxipur, Laxipurin, Modane, NSC 38626, NSC 646568, NSC 7210, Zwitsalax, Dithranol Impurity B
- Formula:C₁₄H₈O₄
- Molecular weight:240.21 g/mol
- Description:Dantron
- Appearance:Neat